Learn More
Claudin-1 Antibody (CL3698) [DyLight 680], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61630FR
Product Code. 30305130
  • € 737.00 / 0.10mL
Description
Claudin-1 Monoclonal antibody specifically detects Claudin-1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Claudin-1
Monoclonal
DyLight 680
claudin 1, claudin-1, CLD1, ILVASCSenescence-associated epithelial membrane protein, SEMP1senescence-associated epithelial membrane protein 1
This antibody was developed using a recombinant protein derived from O95832, with the exact immunogen sequence remaining proprietary.
0.1 mL
Extracellular Matrix
9076
Store at 4⪚C in the dark.
IgG1
Western Blot
CL3698
50mM Sodium Borate
Mouse
Protein A purified