missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-77036CL1
This item is not returnable.
View return policy
Description
Claudin-1 Polyclonal antibody specifically detects Claudin-1 in Canine,Human,Mouse,Porcine,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| Claudin-1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Canine, Human, Mouse, Pig, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| claudin 1, claudin-1, CLD1, ILVASCSenescence-associated epithelial membrane protein, SEMP1senescence-associated epithelial membrane protein 1 | |
| Antibody was raised against a peptide corresponding to 20 amino acids near the carboxy terminus of human Claudin-1. The immunogen is located within the last 50 amino acids of Claudin-1. Amino Acid Sequence: PRKTTSYPTPRPYPKPAPS | |
| 0.1 mL | |
| Extracellular Matrix | |
| 9076 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction