missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-10b Antibody (394513) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4279JF635
This item is not returnable.
View return policy
Description
Claudin-10b Monoclonal antibody specifically detects Claudin-10b in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| Claudin-10b | |
| Monoclonal | |
| Janelia Fluor 635 | |
| CLDN10;CPETRL3;HELIX;OSPL;OSP-L | |
| NS0 mouse myeloma cell line transfected with human Claudin-10 b Isoform, Met1-Val228, Accession # P78369 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 394513 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 9071 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction