Learn More
Claudin-12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-87450
This item is not returnable.
View return policy
Description
Claudin-12 Polyclonal specifically detects Claudin-12 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Claudin-12 | |
| Polyclonal | |
| CLDN12 | |
| Affinity Purified | |
| IgG |
| Western Blot | |
| claudin 12, claudin-12 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:SLPSPFWQPLYSHPPSMHTYSQPYSARSRLSAIEIDIPVVSHTT | |
| 0.1 mL |