Learn More
Claudin-12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand:  Novus Biologicals NBP1-87450
 View more versions of this product
Product Code. 18756753
  • € 539.00 / 0.10mL
Description
Claudin-12 Polyclonal specifically detects Claudin-12 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Claudin-12
Polyclonal
CLDN12
Affinity Purified
IgG
Western Blot
claudin 12, claudin-12
This antibody was developed against Recombinant Protein corresponding to amino acids:SLPSPFWQPLYSHPPSMHTYSQPYSARSRLSAIEIDIPVVSHTT
0.1 mL