missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Claudin 12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ 388200
This item is not returnable.
View return policy
Description
Claudin 12 Polyclonal Antibody for Western Blot, ICC/IF
The claudin superfamily consists of many structurally related proteins in humans. These proteins, which include claudin 1 through 18, are important structural and functional components of tight junctions in paracellular transport. Claudins are located in both epithelial and endothelial cells in all tight junction-bearing tissues. Claudins, which consist of four trans-membrane domains and two extracellular loops, make up tight junction strands.
Specifications
| Claudin 12 | |
| Polyclonal | |
| Unconjugated | |
| CLDN12 | |
| claudin 12; claudin-12; Cldn12; RGD1561053 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 500000, 64945, 9069 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.25 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| P56749, Q9ET43 | |
| CLDN12 | |
| Synthetic peptide derived from the N-terminal region of mouse and human Claudin-12. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction