missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Claudin 14 Polyclonal Antibody
GREENER_CHOICE

Product Code. 11558641
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11558641 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11558641 Supplier Invitrogen™ Supplier No. PA19540

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chicken Polyclonal Antibody

PA1-9540 detects CLDN14 in human samples. PA1-9540 has been successfully used in Western blot procedures. By Western blot, PA1-9540 detects a 26 kDa protein representing CLDN14. PA1-9540 immunogen is a purified recombinant CLDN14 corresponding to the chromosome 21 gene (Human).

Tight junctions represent one mode of cell-to-cell adhesion in epithelial or endothelial cell sheets, forming continuous seals around cells and serving as a physical barrier to prevent solutes and water from passing freely through the paracellular space. These junctions are comprised of sets of continuous networking strands in the outwardly facing cytoplasmic leaflet, with complementary grooves in the inwardly facing extracytoplasmic leaflet. The protein encoded by this gene, a member of the claudin family, is an integral membrane protein and a component of tight junction strands. The encoded protein also binds specifically to the WW domain of Yes-associated protein. Defects in this gene are the cause of an autosomal recessive form of nonsyndromic sensorineural deafness. Several transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Claudin 14
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CLDN14
Gene Accession No. O95500
Gene Alias AI851731; claudin 14; claudin-14; CLDN14; DFNB29; UNQ777/PRO1571
Gene Symbols CLDN14
Host Species Chicken
Immunogen Purified recombinant CLDN14 corresponding to the chromosome 21 gene (Human).
Purification Method IgY fraction
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23562
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.