missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-15 Antibody (540315) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10344JF585
This item is not returnable.
View return policy
Description
Claudin-15 Monoclonal antibody specifically detects Claudin-15 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| Claudin-15 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| claudin 15, claudin-15, FLJ42715, MGC19536 | |
| NS0 mouse myeloma cell line transfected with human Claudin-15, Accession # P56746 | |
| RUO | |
| 24146 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 540315 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction