missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-17 Antibody (410509), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4619B
This item is not returnable.
View return policy
Description
Claudin-17 Monoclonal antibody specifically detects Claudin-17 in Human samples. It is validated for Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| Claudin-17 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 26285 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOF | |
| 410509 | |
| Flow Cytometry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| claudin 17, human CLDN17 gene for claudin-17, 10claudin-17, MGC126552, MGC126554 | |
| NS0 mouse myeloma cell line transfected with human Claudin-17, Met1-Val224, Accession # P56750 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction