Learn More
Claudin-17 Antibody (410509), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB4619MFV610
Product Code. 18529759
  • € 618.00 / 0.10mL
Description
Claudin-17 Monoclonal antibody specifically detects Claudin-17 in Human samples. It is validated for Flow Cytometry, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Claudin-17
Monoclonal
mFluor Violet 610 SE
50mM Sodium Borate
Mouse
0.1 mL
Primary
Human
IgG2b
Flow Cytometry, CyTOF
410509
Flow Cytometry, Intracellular Staining by Flow Cytometry, CyTOF-ready
claudin 17, human CLDN17 gene for claudin-17, 10claudin-17, MGC126552, MGC126554
NS0 mouse myeloma cell line transfected with human Claudin-17, Met1-Val224, Accession # P56750
RUO
26285
Store at 4C in the dark.