missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-4 Antibody (2G12), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 343.00 - € 533.00
Specifications
| Antigen | Claudin-4 |
|---|---|
| Clone | 2G12 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043785
|
Novus Biologicals
NBP3-26655-50ul |
50 μL |
€ 343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043398
|
Novus Biologicals
NBP3-26655-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Claudin-4 Monoclonal antibody specifically detects Claudin-4 in Human samples. It is validated for ELISASpecifications
| Claudin-4 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| claudin 4, Clostridium perfringens enterotoxin receptor, Clostridium perfringens enterotoxin receptor 1, CPE-RCPETR, CPE-receptor, hCPE-R, WBSCR8CPERCPETR1claudin-4, Williams-Beuren syndrome chromosomal region 8 protein | |
| A synthesized peptide derived from Human Claudin-4 [UniProt O14493] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 2G12 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Cancer, Extracellular Matrix | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 1364 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title