missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-4 Antibody (382321), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4219C
This item is not returnable.
View return policy
Description
Claudin-4 Monoclonal antibody specifically detects Claudin-4 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Claudin-4 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 382321 | |
| Flow Cytometry, CyTOF-ready | |
| claudin 4, Clostridium perfringens enterotoxin receptor, Clostridium perfringens enterotoxin receptor 1, CPE-RCPETR, CPE-receptor, hCPE-R, WBSCR8CPERCPETR1claudin-4, Williams-Beuren syndrome chromosomal region 8 protein | |
| NS0 mouse myeloma cell line transfected with human Claudin-4, Met1-Val209, Accession # NP_001296 | |
| 0.1 mL | |
| Cancer, Extracellular Matrix | |
| 1364 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction