Learn More
Claudin-5 Antibody (OTI1G4), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-71329V
Product Code. 18153283
  • ONLINE EXCLUSIVE PRICE
    € 795.00
    € 751.80 / 0.10mL
    Save € 43.20
    5% Off
Description
Claudin-5 Monoclonal specifically detects Claudin-5 in Human, Mouse, Rat, Porcine samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Claudin-5
Monoclonal
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
CLDN5
Full length recombinant protein of human Claudin-5 (NP_003268) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat, Pig
IgG1
Flow Cytometry, Immunocytochemistry, Immunofluorescence
OTI1G4
Flow Cytometry, Immunocytochemistry/Immunofluorescence
AWALTransmembrane protein deleted in VCFS, BEC1, claudin 5, claudin-5, CPETRL1, TMVCFTMDVCF, transmembrane protein deleted in velocardiofacial syndrome
Mouse
Affinity Purified
Cellular Markers
7122.0
Store at 4°C in the dark.