missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Claudin 6/9 Recombinant Mouse Monoclonal Antibody (9E2)

Mouse Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560736
This item is not returnable.
View return policy
Description
Claudin 6/9 Recombinant Monoclonal Antibody for Flow, ELISA
Plays a major role in tight junction-specific obliteration of the intercellular space.
Specifications
| Claudin 6/9 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P56747 | |
| CLDN6 | |
| Recombinant Human CLDN6/9 protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a Fc |
| ELISA, Flow Cytometry | |
| 9E2 | |
| Unconjugated | |
| CLDN6 | |
| Claudin6; Claudin-6; CLD6_HUMAN; CLDN 6; CLDN6; OTTHUMP00000159248; Skullin; Skullin 2; UNQ757/PRO1488 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9074 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction