missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-6 Antibody (342927), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3656PECY7
This item is not returnable.
View return policy
Description
Claudin-6 Monoclonal antibody specifically detects Claudin-6 in Human samples. It is validated for Flow Cytometry
Specifications
| Claudin-6 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 342927 | |
| Flow Cytometry | |
| claudin 6, claudin-6, skullin | |
| NS0 mouse myeloma cell line transfected with human Claudin-6, Met1-Val220, Accession # P56747 | |
| 0.1 mL | |
| Stem Cell Markers | |
| 9074 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction