missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ NAT13 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30133
This item is not returnable.
View return policy
Description
NAT13 Polyclonal antibody specifically detects NAT13 in Human samples. It is validated for Immunoprecipitation
Specifications
| NAT13 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| Unconjugated | |
| EC 2.3.1.-, FLJ13194, hSAN, Mak3 homolog (S. cerevisiae), N(alpha)-acetyltransferase 50, NatE catalytic subunit, N-acetyltransferase 13 (GCN5-related), N-acetyltransferase 5, N-acetyltransferase san homolog, NAT13San, NAT5SAN, NatE catalytic subunit, separation anxiety | |
| Between 119 and 169 | |
| 100 μg | |
| Cell Biology | |
| 80218 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction