Learn More
Novus Biologicals™ Claudin-6 Antibody (IMAB027) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28797-1mg
 View more versions of this product
Product Code. 30046858
  • € 920.00 / 1mg
Description
Claudin-6 Monoclonal antibody specifically detects Claudin-6 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Claudin-6
Monoclonal
Unconjugated
P56747
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
IMAB027
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
claudin 6, claudin-6, skullin
CLDN6
1 mg
Stem Cell Markers
9074