missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-6 Antibody (OTI2H1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71416CL1
This item is not returnable.
View return policy
Description
Claudin-6 Monoclonal antibody specifically detects Claudin-6 in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| Claudin-6 | |
| Monoclonal | |
| CoraFluor 1 | |
| claudin 6, claudin-6, skullin | |
| Full length human recombinant protein of human CLDN6(NP_067018) produced in HEK293T cell. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 9074 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI2H1 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction