missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Claudin 6 Recombinant Mouse Monoclonal Antibody (8D7)

Mouse Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560735
This item is not returnable.
View return policy
Description
Claudin 6 Recombinant Monoclonal Antibody for Flow, ELISA
Tight junctions represent one mode of cell-to-cell adhesion in epithelial or endothelial cell sheets, forming continuous seals around cells and serving as a physical barrier to prevent solutes and water from passing freely through the paracellular space. These junctions are comprised of sets of continuous networking strands in the outwardly facing cytoplasmic leaflet, with complementary grooves in the inwardly facing extracytoplasmic leaflet. This gene encodes a component of tight junction strands, which is a member of the claudin family. The protein is an integral membrane protein and is one of the entry cofactors for hepatitis C virus. The gene methylation may be involved in esophageal tumorigenesis. This gene is adjacent to another family member CLDN9 on chromosome 16.
Specifications
| Claudin 6 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P56747 | |
| CLDN6 | |
| Recombinant Human CLDN6 protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a Fc |
| ELISA, Flow Cytometry | |
| 8D7 | |
| Unconjugated | |
| CLDN6 | |
| AL024037; claudin 6; Claudin6; claudin-6; CLD6; CLDN6; Skullin; tight junction molecule claudin 6; UNQ757/PRO1488 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9074 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction