missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-7 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18508G
This item is not returnable.
View return policy
Description
Claudin-7 Polyclonal antibody specifically detects Claudin-7 in Human samples. It is validated for Western Blot
Specifications
| Claudin-7 | |
| Polyclonal | |
| Western Blot | |
| CEPTRL2, claudin 7, claudin-1, claudin-7, CLDN-7, clostridium perfringens enterotoxin receptor-like 2, CPETRL2 | |
| Recognizes region between residues Between between 161 and 211.. | |
| 0.1 mL | |
| Breast Cancer, Cancer | |
| 1366 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction