missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Claudin 8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ 400700Z
This item is not returnable.
View return policy
Description
Claudin 8 Polyclonal Antibody for Western Blot, IHC (F), ICC/IF
Claudin-8 belongs to the claudin family of integral membrane proteins. Claudins are responsible for the formation of tight junction strands, and are connected with the actin cytoskeleton mediated by ZO-1. Claudin-8 is a tight junction protein expressed in several distal nephron segments, suggesting that it may have a role as a paracellular cation barrier protein. Claudin-8 is expressed in the inner ear, and the umbrella cell tight junctions in the bladder epithelium.
Specifications
| Claudin 8 | |
| Polyclonal | |
| Unconjugated | |
| CLDN8 | |
| AI648025; claudin 8; claudin-8; CLDN8; epididymis secretory protein Li 79; HEL-S-79; UNQ779/PRO1573 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 304124, 478401, 54420 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry | |
| 0.25 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| Q9Z260 | |
| CLDN8 | |
| Synthetic peptide derived from the C-terminal region of mouse and rat Claudin-8. | |
| 100 μg | |
| Primary | |
| Canine, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction