Learn More
Invitrogen™ Claudin 9 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5104427
Product Code. 16680875
  • € 487.00 / 100µL
Description
Antibody detects endogenous levels of total Claudin 9.
CLDN9 is a member of claudin family. It is a four-transmembrane protein with WWCC motif, defined by W-X(17-22)-W-X(2)-C-X(8-10)-C.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Claudin 9
Polyclonal
Unconjugated
Cldn9
claudin 9; claudin-9; CLDN9; nmf329
Rabbit
Affinity chromatography
RUO
287099, 56863, 9080
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O95484, Q9Z0S7
Cldn9
A synthesized peptide derived from human CLDN9(Accession O95484), corresponding to amino acid residues G179-V217.
100 μL
Primary
Human, Mouse, Rat