missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Clavesin-1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140190
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: QSVVEAGTLK HEEKGENENT QPLLALD Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.
Required for normal morphology of late endosomes and/or lysosomes in neurons. Binds phosphatidylinositol 3,5-bisphosphate (PtdIns(3,5)P2).
Specifications
| Clavesin-1 | |
| Polyclonal | |
| Unconjugated | |
| CLVS1 | |
| Cellular retinaldehyde-binding protein-like; clathrin vesicle-associated Sec14 protein 1; clavesin 1; clavesin-1; CLVS1; CRALBPL; retinaldehyde binding protein 1-like 1; retinaldehyde-binding protein 1-like 1; RLBP1L1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 157807 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IUQ0 | |
| CLVS1 | |
| Recombinant protein corresponding to Human CLVS1. Recombinant protein control fragment (Product #RP-109575). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction