missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLCA1 Monoclonal Antibody (OTI8E7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527222
This item is not returnable.
View return policy
Description
CLCA1 Monoclonal Antibody for Western Blot, IHC (P)
This gene encodes a member of the calcium sensitive chloride conductance protein family. To date, all members of this gene family map to the same region on chromosome 1p31-p22 and share a high degree of homology in size, sequence, and predicted structure, but differ significantly in their tissue distributions. The encoded protein is expressed as a precursor protein that is processed into two cell-surface-associated subunits, although the site at which the precursor is cleaved has not been precisely determined. The encoded protein may be involved in mediating calcium-activated chloride conductance in the intestine.
Specifications
| CLCA1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A8K7I4 | |
| CLCA1 | |
| Human recombinant protein fragment corresponding to amino acids 344-412 of CLCA1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI8E7 | |
| Unconjugated | |
| CLCA1 | |
| CACC; CACC1; CaCC-1; Calcium-activated chloride channel family member 1; calcium-activated chloride channel family member 3; Calcium-activated chloride channel protein 1; calcium-activated chloride channel regulator 1; calcium-dependent chloride channel-1; chloride channel accessory 1; chloride channel calcium activated 1; chloride channel calcium activated 2; chloride channel calcium activated 3; chloride channel regulator 1; chloride channel, calcium activated, family member 1; CLCA family member 1, chloride channel regulator; Clca1; Clca2; Clca3; CLCRG1; Gob5; gob-5; hCaCC-1; hCLCA1; mCLCA3; Protein gob-5 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1179 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu