missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLCA4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550209
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total CLCA4 protein.
The protein encoded by this gene belongs to the calcium sensitive chloride conductance protein family. To date, all members of this gene family map to the same site on chromosome 1p31-p22 and share high degrees of homology in size, sequence and predicted structure, but differ significantly in their tissue distributions. Alternative splicing results in multiple transcript variants, only one of which is thought to be protein coding.
Specifications
| CLCA4 | |
| Polyclonal | |
| Unconjugated | |
| CLCA4 | |
| CaCC; CaCC2; CaCC-2; Calcium-activated chloride channel family member 4; calcium-activated chloride channel protein 2; Calcium-activated chloride channel regulator 4; Calcium-activated chloride channel regulator 4, 110 kDa form; Calcium-activated chloride channel regulator 4, 30 kDa form; Chloride channel accessory 4; chloride channel regulator 4; chloride channel, calcium activated, family member 4; CLCA4; hCaCC-2; hCLCA4; UNQ562/PRO1124 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22802 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q14CN2 | |
| CLCA4 | |
| Fusion protein corresponding to a region derived from internal residues of human Calcium-activated chloride channel regulator 4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction