missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLCN2 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97349AF647
This item is not returnable.
View return policy
Description
CLCN2 Polyclonal specifically detects CLCN2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| CLCN2 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| chloride channel 2, chloride channel protein 2, CIC-2, clC-2, CLC2, ECA2, ECA3, EGI11, EGI3, EGMA, EJM6, EJM8 | |
| Produced in rabbits immunized with E. coli-derived Human CLCN2 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 1181 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction