missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLCN2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92467-0.02ml
This item is not returnable.
View return policy
Description
CLCN2 Polyclonal antibody specifically detects CLCN2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| CLCN2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| chloride channel 2, chloride channel protein 2, CIC-2, clC-2, CLC2, ECA2, ECA3, EGI11, EGI3, EGMA, EJM6, EJM8 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 550-770 of human CLCN2 (NP_001164560.1). PSLYDSIIRIKKLPYLPELGWGRHQQYRVRVEDIMVRDVPHVALSCTFRDLRLALHRTKGRMLALVESPESMILLGSIERSQVVALLGAQLSPARRRQHMQERRATQTSPLSDQEGPPTPEASVCFQVNTEDSAFPAARGETHKPLKPALKRGPSVTRNLGESPTGSAESAGIALRSLFCGSPPPEAASEKLESCEKRKLKRVRISLASDADLEGEMSPEE | |
| 0.02 mL | |
| Cancer, Endocrinology, Neuroscience, Signal Transduction | |
| 1181 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction