Learn More
Invitrogen™ CLCN2 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA519297
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, canine, mouse, rabbit and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
The transmembrane protein encoded by this gene is a voltage-gated chloride channel that maintains chloride ion homeostasis in various cells. Defects in this gene may be a cause of certain epilepsies. Four transcript variants encoding different isoforms have been found for this gene.
Specifications
| CLCN2 | |
| Polyclonal | |
| Unconjugated | |
| CLCN2 | |
| AL118368; chloride channel 2; chloride channel protein 2; chloride channel, voltage-sensitive 2; chloride voltage-gated channel 2; CIC 2; CIC2; CIC-2; Clc2; ClC-2; CLCN2; ECA2; ECA3; EG13; EGI11; EGI3; EGMA; EJM6; EJM8; LKPAT; nmf240 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 1181 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P51788 | |
| CLCN2 | |
| Synthetic peptide sequence (QQLDEPVNFSDCK) corresponding to the internal amino acids of CLCN2 (aa 777-790). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |