missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLCN3 Antibody (S258-5), Novus Biologicals™
Mouse Monoclonal Antibody
€ 179.00 - € 579.00
Specifications
| Antigen | CLCN3 |
|---|---|
| Clone | S258-5 |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18202492
|
Novus Biologicals
NBP2-59379 |
100 μg |
€ 579.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18691037
|
Novus Biologicals
NBP2-59379-25ug |
25 μg |
€ 179.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CLCN3 Monoclonal specifically detects CLCN3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| CLCN3 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| 1182 | |
| Synthetic peptide amino acids 98-115 (cytoplasmic N-terminus) of rat Clcn3 | |
| Primary | |
| Detects 90kDa. Does not cross-react with Clcn4 or Clcn5 (based on KO validation results). |
| S258-5 | |
| Monoclonal | |
| Purified | |
| RUO | |
| chloride channel 3, Chloride channel protein 3, Chloride transporter ClC-3, ClC-3, CLC3, H(+)/Cl(-) exchange transporter 3 | |
| CLCN3 | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title