Learn More
Invitrogen™ CLCN3 Monoclonal Antibody (N258/5), PE

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545557
Description
1 μg/mL of MA5-45557 was sufficient for detection of Clcn3 in 20 μg of rat brain membrane lysate and assayed by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 90kDa. Does not cross-react with Clcn4 or Clcn5 (based on KO validation results). This antibody was formerly sold as clone S258-5.
Mediates the exchange of chloride ions against protons. Functions as antiporter and contributes to the acidification of the endosome and synaptic vesicle lumen, and may thereby affect vesicle trafficking and exocytosis. May play an important role in neuronal cell function through regulation of membrane excitability by protein kinase C. It could help neuronal cells to establish short-term memory.
Specifications
| CLCN3 | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
| P51790, P51791, P51792 | |
| CLCN3 | |
| Synthetic peptide amino acids 98-115 (cytoplasmic N-terminus) of rat Clcn3. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| N258/5 | |
| PE | |
| CLCN3 | |
| chloride channel 3; chloride channel protein 3; chloride channel, voltage-sensitive 3; chloride transporter ClC-3; chloride voltage-gated channel 3; CIC3; Clc3; clC-3; CLCN 3; Clcn3; DKFZp564I0463; H(+)/Cl(-) exchange transporter 3; protein kinase C-regulated chloride channel | |
| Mouse | |
| Protein G | |
| RUO | |
| 1182, 12725, 84360 | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.