missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLDN3 Mouse anti-Human, Clone: OTI1E7, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806232S
This item is not returnable.
View return policy
Description
CLDN3 Monoclonal antibody specifically detects CLDN3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CLDN3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O15551 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1365 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1E7 | |
| Unconjugated | |
| CLDN3 | |
| AI182374; C7orf1; claudin 3; claudin-3; CLDN3; clostridium perfringens enterotoxin receptor 2; CPE-R 2; CPE-R2; CPE-receptor 2; CPETR2; hRVP1; integral membrane protein claudin-3; mRVP1; Rat ventral prostate.1 protein; Rat ventral prostate.1 protein homolog; RVP1; tight junction protein; ventral prostate.1 protein; ventral prostate.1 protein homolog; ventral prostate.1-like protein | |
| Full length human recombinant protein of human CLDN3 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction