missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLDN4, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00001364-B01.50uL
This item is not returnable.
View return policy
Description
Sequence: MASMGLQVMGIALAVLGWLAVMLCCALPMWRVTAFIGSNIVTSQTIWEGLWMNCVVQSTGQMQCKVYDSLLALPQDLQAARALVIISIIVAALGVLLSVVGGKCTNCLEDESAKAKTMIVAGVVFLLAGLMVIVPVSWTAHNIIQDFYNPLVASGQKREMGASLYVGWAASGLLLLGGGLLCCNCPPRTDKPYSAKYSAARSAAASNYVSpecifications
| CLDN4 | |
| Polyclonal | |
| No additive | |
| NM_001305.3 | |
| CLDN4 | |
| CLDN4 (NP_001296.1, 1 a.a. ∼ 209 a.a) full-length human protein. | |
| RUO | |
| 1364 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| claudin 4 | |
| CPE-R/CPER/CPETR/CPETR1/WBSCR8/hCPE-R | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction