Learn More
CLDN6 Mouse anti-Human, Clone: OTI4B5, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF506897
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Tight junctions represent one mode of cell-to-cell adhesion in epithelial or endothelial cell sheets, forming continuous seals around cells and serving as a physical barrier to prevent solutes and water from passing freely through the paracellular space. These junctions are comprised of sets of continuous networking strands in the outwardly facing cytoplasmic leaflet, with complementary grooves in the inwardly facing extracytoplasmic leaflet. This gene encodes a component of tight junction strands, which is a member of the claudin family. The protein is an integral membrane protein and is one of the entry cofactors for hepatitis C virus. The gene methylation may be involved in esophageal tumorigenesis. This gene is adjacent to another family member CLDN9 on chromosome 16.Specifications
| CLDN6 | |
| Monoclonal | |
| Unconjugated | |
| CLDN6 | |
| claudin 6 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9074 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI4B5 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P56747 | |
| CLDN6 | |
| Full length human recombit protein of human CLDN6 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.