missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLDND2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551345
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total CLDND2 protein.
CLDND2 (claudin domain containing 2) is a 167 amino acid multi-pass membrane protein that belongs to the PMP-22/EMP/MP20 family and is encoded by a gene that maps to human chromosome 19q13.33. Consisting of around 63 million bases with over 1,400 genes, chromosome 19 makes up over 2% of human genomic DNA. Chromosome 19 includes a diversity of interesting genes and is recognized for having the greatest gene density of the human chromosomes.
Specifications
| CLDND2 | |
| Polyclonal | |
| Unconjugated | |
| CLDND2 | |
| 1700071E18Rik; claudin domain containing 2; claudin domain-containing protein 2; CLDN2; CLDND2; MGC33839 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 125875, 74276 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 3 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q8NHS1, Q9D9H2 | |
| CLDND2 | |
| Synthetic peptide of human CLDND2. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction