Learn More
Cole-Parmer Cleanroom Hose Barb Fittings, Tee Union, Natural Kynar™
Product Code. p-200105112 Shop All Cole-Parmer Products
Click to view available options
Diameter (English) Inner:
1 in.
1/16 in.
1/2 in.
1/4 in.
1/8 in.
3/16 in.
3/32 in.
3/4 in.
3/8 in.
5/16 in.
5/32 in.
5/8 in.
Unit Size:
Pack of 10
Product Code. 30053835
Brand: Cole-Parmer 5011095
Only null left
Add to basket
Add to basket
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Quick and easy assembly without tools. Single barb design tightly grips the tubing. Biopharma compatible and USP 661 and BPOG compliant
Specifications
Certifications/Compliance USP;Non-DEHP;Non-Phthalate
Diameter (English) Inner 5/32 in.
Min. Operating Temperature (Metric) -35°C
Packaging Quantity 10/Pk.
Outlet Port Hose Barb Tee
Sterilization Method E-beam/gamma irradiation, EtO, or autoclave up to 135°C
Shape Tee
Color Natural