Learn More
Cole-Parmer Cleanroom Hose Barb Fittings, Y Union, White Nylon
Product Code. 30053512 Shop All Cole-Parmer Products
Click to view available options
Diameter (English) Inner:
1/16 in.
1/2 in.
1/4 in.
1/8 in.
3/16 in.
3/32 in.
3/8 in.
5/8 in.
Unit Size:
Pack of 10
Product Code. 30053512
Brand: Cole-Parmer 5011118
Only null left
Add to basket
Add to basket
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Quick and easy assembly without tools. Single barb design tightly grips the tubing. FDA and RoHS compliant
Specifications
Certifications/Compliance FDA;Non-DEHP;Non-Phthalate;RoHS
Diameter (English) Inner 3/32 in.
Packaging Quantity 10/Pk.
Outlet Port Hose Barb Y
Sterilization Method E-beam/gamma irradiation, EtO, or autoclave up to 121°C
Shape Y
Color White