Learn More
Cole-Parmer Cleanroom Male Slip Luer to Barb Fittings, Straight Adapter, White Nylon
Shop All Cole-Parmer Products
Click to view available options
Diameter (English) Inner:
1/16 in.
1/4 in.
1/8 in.
3/16 in.
3/32 in.
5/32 in.
Unit Size:
Pack of 10
Description
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Connect to a female luer body to form a leak-free fluid path. Single barb design tightly grips the tubing. Meets FDA regulations 21 CFR 178.3297 and RoHS compliant
Specifications
Specifications
| Certifications/Compliance | FDA;Non-DEHP;Non-Phthalate;RoHS |
| Diameter (English) Inner | 1/4 in. |
| Material | Nylon |
| Packaging Quantity | 10/Pk. |
| Inlet | Male slip luer |
| Color | White |