Learn More
Cole-Parmer Cleanroom Male Slip Luer Fittings, Elbow Union
Product Code. 30053046 Shop All Cole-Parmer Products
Click to view available options
Material:
CrystalVu (CVU)
Kynar
Nylon
Polypropylene
Color:
Clear
Natural
White
Unit Size:
Pack of 10
Product Code. 30053046
Brand: Cole-Parmer 5011543
Only null left
Add to basket
Add to basket
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Connect to female luer bodies to form a leak-free fluid path
Specifications
Certifications/Compliance Non-DEHP;Non-Phthalate;RoHS;USP
Material CrystalVu (CVU)
Packaging Quantity 10/Pk.
Inlet Male Slip Luer
Color Clear
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.