Learn More
Hirschmann™ Clear Glass Graduated Pipettes with Amber Stain Graduation
Description
- Short line
- Acc. to DIN EN ISO 835
- Graduated to tip, zero on top, type 3
Specifications
Specifications
| Class | B |
| Material | Soda Lime Glass |
| Graduated | Yes |
| Graduation Color | Amber |
| Graduations | 0.1 mL |
| Graduation Type | Amber graduations permanently with silver diffusion color |
| Quantity | 12 Pack |
| Volume (Metric) | 5 mL |
| Length (Metric) | 360 ±5 mm |
| Type | Graduated Pipet |