Learn More
Berlin Packaging™ Clear Laboratory Glassware Pot
Product Code. 10207994
Change view
Click to view available options
Capacity (Metric):
100 mL
150 mL
250 mL
370 mL
500 mL
1000 mL
Quantity:
12 Set
18 Set
20 Set
42 Set
48 Set
63 Set
72 Set
Unit Size:
Pack of 12
Pack of 18
Pack of 20
Pack of 2688
Pack of 4158
Pack of 42
Pack of 48
Pack of 63
Pack of 72
Pack of 720
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Quantity unitSize
10207994 500 mL 20 Set Pack of 20
10554375 1000 mL 18 Set Pack of 18
30408780 1000 mL 18 Set Pack of 720
10647075 100 mL 63 Set Pack of 63
30408786 100 mL 63 Set Pack of 4158
10744347 150 mL 42 Set Pack of 42
10195884 250 mL 48 Set Pack of 48
30408783 250 mL 48 Set Pack of 2688
10647845 100 mL 72 Set Pack of 72
10310015 370 mL 12 Set Pack of 12
30408784 500 mL 20 Set Pack of 720
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 10207994 Supplier Berlin Packaging™ Supplier No. 1000716
Add to basket
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Use the Berlin Packaging™ Clear Laboratory Glassware Pot as a fundamental piece of laboratory glassware. The autoclavable clear glass is breakproof and chemically and thermally shockproof.
  • Clear borosilicate glass
  • Break-resistant
  • Chemically and thermally shockproof;
  • In compliance with EP, USP and JP guidelines
  • Autoclavable
Specifications
Certifications/Compliance DIN 55
Quantity 20 Set
Capacity (Metric) 500 mL
Material Glass
Product Type Pot