Learn More
Alpha Packaging™ Clear Laboratory Glassware Pot
Use the Alpha Packaging™ Clear Laboratory Glassware Pot as a fundamental piece of laboratory glassware. The autoclavable clear glass is breakproof and chemically and thermally shockproof.
Brand: Alpha Packaging™ 231262
Additional Details : Weight : 5.80500kg
Description
- Clear borosilicate glass
- Break-resistant
- Chemically and thermally shockproof;
- In compliance with EP, USP and JP guidelines
- Autoclavable
Specifications
Pot | |
Glass | |
DIN 40 |
63 Set | |
100 mL |