Learn More
CLEC-1 Antibody (MM0187-8K42), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-12192AF700
Product Code. 18092329
  • € 599.00 / 0.10mL
Description
CLEC-1 Monoclonal specifically detects CLEC-1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CLEC-1
Monoclonal
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
CLEC1A
Human recombinant CLEC-1 (extracellular domain)
0.1 mL
51267
Store at 4C in the dark.
IgG2
Western Blot
MM0187-8K42
Western Blot
CLEC-1, CLEC1C-type lectin domain family 1 member A, C-type lectin domain family 1, member A, C-type lectin-like receptor 1, C-type lectin-like receptor-1, MGC34328
Mouse
Protein G purified