missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC-1 Antibody (MM0187-8K42), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12192F
This item is not returnable.
View return policy
Description
CLEC-1 Monoclonal antibody specifically detects CLEC-1 in Human samples. It is validated for Western Blot
Specifications
| CLEC-1 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 51267 | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot | |
| MM0187-8K42 | |
| Western Blot | |
| CLEC-1, CLEC1C-type lectin domain family 1 member A, C-type lectin domain family 1, member A, C-type lectin-like receptor 1, C-type lectin-like receptor-1, MGC34328 | |
| Human recombinant CLEC-1 (extracellular domain) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction