missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC-2/CLEC-1B Antibody (004), DyLight 680 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89727FR
This item is not returnable.
View return policy
Description
CLEC-2/CLEC-1B Monoclonal antibody specifically detects CLEC-2/CLEC-1B in Human samples. It is validated for ELISA
Specifications
| CLEC-2/CLEC-1B | |
| Monoclonal | |
| DyLight 680 | |
| 1810061I13Rik, CLEC-2, CLEC2B, CLEC2PRO1384, C-type lectin domain family 1 member B, C-type lectin domain family 1, member B, C-type lectin-like receptor 2, C-type lectin-like receptor-2, QDED721 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CLEC-2/CLEC-1B (Gln 58-Pro 229; NP_057593.3). | |
| 0.1 mL | |
| Immunology, Innate Immunity, Signal Transduction | |
| 51266 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 004 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction