Learn More
CLEC-2/CLEC-1B Antibody (AYP1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50166-0.025ml
 View more versions of this product
Product Code. 18697906
  • € 201.00 / 0.03mL
Description
CLEC-2/CLEC-1B Monoclonal antibody specifically detects CLEC-2/CLEC-1B in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Functional Assay, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CLEC-2/CLEC-1B
Monoclonal
1 mg/mL
Flow Cytometry 1:10 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Functional, CyTOF-ready
1810061I13Rik, CLEC-2, CLEC2B, CLEC2PRO1384, C-type lectin domain family 1 member B, C-type lectin domain family 1, member B, C-type lectin-like receptor 2, C-type lectin-like receptor-2, QDED721
A recombinant extracellular fragment of human CLEC-2/CLEC-1B (aa 68-229), which is the extracellular domain of the protein
0.025 mL
Immunology, Innate Immunity, Signal Transduction
51266
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Functional Assay, CyTOF
AYP1
Unconjugated
PBS
Mouse
Protein A purified