missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC-2/CLEC-1B Antibody (AYP2), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50167MFV610
This item is not returnable.
View return policy
Description
CLEC-2/CLEC-1B Monoclonal antibody specifically detects CLEC-2/CLEC-1B in Human samples. It is validated for Western Blot
Specifications
| CLEC-2/CLEC-1B | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| AYP2 | |
| Western Blot | |
| 1810061I13Rik, CLEC-2, CLEC2B, CLEC2PRO1384, C-type lectin domain family 1 member B, C-type lectin domain family 1, member B, C-type lectin-like receptor 2, C-type lectin-like receptor-2, QDED721 | |
| A recombinant extracellular fragment of human CLEC-2/CLEC-1B (aa 68-229), which is the extracellular domain of the protein. | |
| 0.1 mL | |
| Immunology, Innate Immunity, Signal Transduction | |
| 51266 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction