missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC16A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
€ 433.00 - € 539.00
Specifications
| Antigen | CLEC16A |
|---|---|
| Dilution | Western Blot Reported in scientific literature (PMID: 22002632)., Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18470751
|
Novus Biologicals
NBP1-81805-25ul |
25 μL |
€ 433.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18769373
|
Novus Biologicals
NBP1-81805 |
€ 539.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
CLEC16A Polyclonal specifically detects CLEC16A in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CLEC16A | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Rat | |
| Q2KHT3 | |
| 23274 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:VPYFSNLVWFIGSHVIELDDCVQTDEEHRNRGKLSDLVAEHLDHLHYLNDILIINCEFLNDVLTDHLLNRLFLPLYVYSLENQDKGGERPKISLPVSLYLLSQVFL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot Reported in scientific literature (PMID: 22002632)., Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Immunology, Innate Immunity | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| C-type lectin domain family 16, member A, KIAA0350Gop-1, MGC111457, protein CLEC16A | |
| CLEC16A | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title