missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLEC17A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548997
This item is not returnable.
View return policy
Description
CLEC17A Polyclonal Antibody for Western Blot
Cell surface receptor which may be involved in carbohydrate-mediated communication between cells in the germinal center. Binds glycans with terminal alpha-linked mannose or fucose residues.
Specifications
| CLEC17A | |
| Polyclonal | |
| Unconjugated | |
| CLEC17A | |
| CLEC17A; C-type lectin domain family 17 member A; C-type lectin domain family 17, member A; Prolectin | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 388512 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.3 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6ZS10 | |
| CLEC17A | |
| KLH conjugated synthetic peptide between 176-203 amino acids from the C-terminal region of human CLEC17A. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction