missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLEC19A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572162
This item is not returnable.
View return policy
Description
CLEC19A Polyclonal Antibody for Western Blot
CLEC19A (C-Type Lectin Domain Containing 19A) is a Protein Coding gene. Diseases associated with CLEC19A includeTobacco Addiction. Gene Ontology (GO) annotations related to this gene includecarbohydrate binding.
Specifications
| CLEC19A | |
| Polyclonal | |
| Unconjugated | |
| CLEC19A | |
| CLEC19A; C-type lectin domain family 19 member A; C-type lectin domain family 19, member A | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 728276 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.32 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6UXS0 | |
| CLEC19A | |
| KLH conjugated synthetic peptide between 54-80 amino acids from the Central region of human CLEC19A. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction