Learn More
Invitrogen™ CLEC1A Monoclonal Antibody (2F7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524458
Description
Peptide Sequence: MQAKYSSTRD MLDDDGDTTM SLHSQGSATT RHPEPRRTEH RAPSSTWRPV ALTLLTLCLV LLIGLAALGL LFFQYYQLSN TGQDTISQME ERLGNTSQEL QSLQVQNIKL AGSLQHVAEK LCRELYNKAG AHRCSPRTEQ WKWHGDNCYQ FYKDSKSWED CKYFCLSENS TMLKINKQED LEFAASQSYS EFFYSYWTGL LRPDSGKAWL WMDGTPFTSE LFHIIIDVTS PRSRDCVAIL NGMIFSKDCK ELKRCVCERR AGMVKPESLH VPPETLGEGD.
This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded protein may play a role in regulating dendritic cell function. Alternative splice variants have been described but their biological nature has not been determined. This gene is closely linked to other CTL/CTLD superfamily members on chromosome 12p13 in the natural killer gene complex region.
Specifications
| CLEC1A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q8NC01 | |
| CLEC1A | |
| CLEC1A (AAH39072, 1 a.a. ~ 280 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 2F7 | |
| Unconjugated | |
| CLEC1A | |
| 5930406N14Rik; CLEC1; CLEC-1; CLEC1A; C-type lectin domain family 1 member A; C-type lectin domain family 1, member A; C-type lectin-like receptor 1; C-type lectin-like receptor-1; RGD1566110; UNQ569/PRO1131 | |
| Mouse | |
| Protein A | |
| RUO | |
| 51267 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.