missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CLEC1A Polyclonal Antibody
GREENER_CHOICE

Product Code. 15911665
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15911665 100 μg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15911665 Supplier Invitrogen™ Supplier No. PA573008

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Sequence homology: Orangutan, chimpanzee, and white-cheeked gibbon (100%), New world monkey, crab-eating macaque, and rhesus monkey (89%), Porcine (78%), Canine, african bush elephant, and rabbit (72%), panda, cattle, chinese hamster, and horse (67%), Rat (61%).

This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded protein may play a role in regulating dendritic cell function. Alternative splice variants have been described but their biological nature has not been determined. This gene is closely linked to other CTL/CTLD superfamily members on chromosome 12p13 in the natural killer gene complex region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC1A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA and 0.05% sodium azide
Gene CLEC1A
Gene Accession No. Q8BWY2, Q8NC01
Gene Alias 5930406N14Rik; CLEC1; CLEC-1; CLEC1A; C-type lectin domain family 1 member A; C-type lectin domain family 1, member A; C-type lectin-like receptor 1; C-type lectin-like receptor-1; RGD1566110; UNQ569/PRO1131
Gene Symbols CLEC1A
Host Species Rabbit
Immunogen A portion of amino acids 50-100 from human CLEC1.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 243653, 51267
Target Species Human, Mouse, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.